Learn More
Medchemexpress LLC S-Trityl-L-cysteine | 2799-07-7 | 99.0% | 363.47 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW01110250MG
Encompass_Preferred
S-Trityl-L-cysteine is a selective and allosteric kinesin Eg5 inhibitor, demonstrating antitumor activities. It inhibits basal ATPase activity with an IC50 of 1 μM and microtubule-activated ATPase activity with an IC50 of 140 nM. The compound binds to a unique pocket within the Eg5 motor domain.
  • Mediates cell apoptosis and cell cycle arrest in a dose-dependent manner.
  • Triggers activation of mitogen-activated protein kinase and nuclear factor κB signaling pathways.
  • Induces mitotic arrest in HeLa cells with characteristic monoastral spindles.
Catalog No. 50-004-52287