Learn More
TARGETMOL CHEMICALS INC S-Vamicamide 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T35033L1MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (S)-Vamicamide is a novel anticholinergic compound.Vamicamide increases spontaneous locomotor activity in mice at 32 mg/kg or higher (p.o.) and inhibits tonic convulsions in mice at 100 mg/kg.13483-86-1 Purity 99.76%
Catalog No. 50-287-6521