Learn More
Medchemexpress LLC (S)-Veludacigib | 2732902-09-7 | C20H19FN4O3S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY153343A
Encompass_Preferred
(S)-BAY 2965501 is the left-handed isomer of BAY 2965501 (HY-153343). It is a potent and selective diacylglycerol kinase zeta (DGKζ) inhibitor that induces pERK activation and can be used for cancer research.
  • Potent and selective diacylglycerol kinase zeta (DGKζ) inhibitor
  • Induces pERK activation
  • Can be used for the research of cancer
Catalog No. 50-003-74677