Learn More
Medchemexpress LLC (S)-Viloxazine-d5 (hydrochloride) | 1246816-39-6 | 98.2% | 278.79 g/mol | C13H15D5ClNO3 | 5MG

Supplier: Medchemexpress LLC HY125784BS5MG
(S)-Viloxazine-d5 (hydrochloride) is the deuterium-labeled form of (S)-viloxazine supplied as a hydrochloride salt for research use only. It is intended as an isotopically labeled standard or tracer for analytical, pharmacokinetic, and mass spectrometry applications.
- Deuterium-labeled (d5) hydrochloride salt.
- Intended for use as an isotopically labeled internal standard or tracer.
- High reported purity (~98.2%).
- Molecular weight ~278.79 g/mol.
- Soluble in DMSO for analytical preparation.
- Supplied in milligram-scale quantities suitable for analytical workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.