Learn More
Medchemexpress LLC S-ZINC-3573 10mg | 2095596-11-3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11568210MG
Encompass_Preferred
(S)-ZINC-3573 is the inactive enantiomer of ZINC-3573 provided for research use as a control probe in studies of the Mas-related G protein-coupled receptor X2 (MRGPRX2). It shows no activity at MRGPRX2 below 100 μM and is supplied as a high-purity solid suitable for biochemical and cellular assays.
  • Inactive enantiomer control for MRGPRX2 studies.
  • High purity suitable for biochemical and cellular assays.
  • Molecular weight 307.39 g/mol and formula C18H21N5.
  • Provided as a small-scale 10 MG package for laboratory use.
  • Supplied with characterization data to support experimental use.
Catalog No. 50-003-07923