Learn More
Medchemexpress LLC S-ZINC-3573 5mg | 2095596-11-3 | 307.39 | C18H21N5 | 5 MG

Supplier: Medchemexpress LLC HY1156825MG
(S)-ZINC-3573 is the inactive enantiomer of ZINC-3573 provided for research use as an internal control to study Mas-related G-protein-coupled receptors (MRGPR). It is supplied as a high-purity solid with documented solubility and storage recommendations for experimental workflows.
- High purity: 99.89%.
- Molecular formula: C18H21N5; molecular weight: 307.39.
- Solid form with recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Soluble in DMSO (50 mg/mL); in vivo formulation protocols available for aqueous dosing.
- Intended for research use only; not for human therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.