Learn More
Genetex Inc S100 beta Monoclonal Antibody [MSVA-490R] HistoMAX™ (GTX04404) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04404
Encompass_Preferred
The S100 beta antibody [MSVA-490R] HistoMAX™ (GTX04404) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human cerebrum (grey matter) tissue using GTX04404 S100 beta antibody [MSVA-490R] HistoMAX. Strong ubiquitous S100 beta staining, except neirons.
Catalog No. NC3666774