Learn More
Medchemexpress LLC S100A2 Human 100ug
SDP
Supplier:  Medchemexpress LLC HYP72481100UG
Encompass_Preferred
The S100A2 protein is a potential calcium sensor that actively promotes cellular calcium signaling and affects various physiological processes As a homodimer it interacts with TPR-containing proteins including FKBP4 and participates in multiple molecular associations S100A2 Protein Human is the recombinant human-derived S100A2 protein expressed by E coli with tag free
Catalog No. 50-001-83011