Learn More
ABclonal Technology S100A7/Psoriasin Rabbit mAb
SDP
Supplier:  ABclonal Technology A2227120UL
Encompass_Preferred
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs S100 proteins are localized in the cytoplasm and or nucleus of a wide range of cells and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation S100 genes include at least 13 members which are located as a cluster on chromosome 1q21 This protein differs from the other S100 proteins of known structure in its lack of calcium binding ability in one EF-hand at the N-terminus The protein is overexpressed in hyperproliferative skin diseases exhibits antimicrobial activities against bacteria and induces immunomodulatory activities
Catalog No. 50-000-05200