Learn More
Medchemexpress LLC S100A8-S100A9 Hetero 100ug

Supplier: Medchemexpress LLC HYP71076100UG
Recombinant human S100A8-S100A9 heterodimer, His-tagged and supplied as a 0.22 μm-filtered buffered solution. Expressed in E. coli and provided at >95% purity, this calprotectin complex is intended for biochemical and immunological assays studying inflammation and leukocyte function; lot-specific datasheet and COA are available.
- Supplied as a filtered, buffered solution for immediate use.
- Expressed in E. coli with a C-terminal 6xHis tag.
- Purity greater than 95% by reducing SDS-PAGE.
- Includes accession-verified human sequences for both subunits.
- Provided with lot-specific datasheet and certificate of analysis.
- Store at -80°C and avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.