Learn More
Medchemexpress LLC S100A8-S100A9 Hetero 100ug
SDP
Supplier:  Medchemexpress LLC HYP71076A100UG
Encompass_Preferred
Recombinant human S100A8-S100A9 heterodimer with a C-terminal 6xHis tag, expressed in E. coli and supplied as a stabilized solution for research applications. The protein models the native calcium- and zinc-binding heterodimer implicated in inflammatory regulation and is suitable for biochemical characterization, assay development, and cellular studies. For research use only.
  • Purified heterodimer suitable for biochemical assays.
  • C-terminal 6xHis tag enables affinity purification and detection.
  • Expressed in E. coli for consistent recombinant production.
  • Supplied in solution for immediate use in experiments.
  • Human sequence validated against UniProt accession numbers.
Catalog No. 50-001-96596