Learn More
Medchemexpress LLC S100A8-S100A9 Hetero 50ug

Supplier: Medchemexpress LLC HYP71076A50UG
Recombinant human S100A8-S100A9 heterodimer supplied as a His-tagged, sterile-filtered solution for biochemical and cell-based studies. The protein is expressed in Escherichia coli, characterized at >95% purity by reducing SDS-PAGE, and provided in lot-specific buffer formulations; typical size offerings include 50 μg, and material should be stored frozen to preserve activity.
- Recombinant human heterodimer expressed in Escherichia coli.
- C-terminal 6xHis tag for affinity detection or purification.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Supplied as a 0.22 μm filtered solution; buffer is lot specific.
- Suitable for biochemical and cell-based assays involving inflammatory signaling.
- Store at -80°C and avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.