Learn More
Medchemexpress LLC S100A9 Human His 50ug
SDP
Supplier:  Medchemexpress LLC HYP70532A50UG
Encompass_Preferred
Recombinant human S100A9 (His-tag) is a lyophilized, E. coli-expressed protein for research into inflammation, neutrophil function, and immune signaling. Supplied as a 50 μg vial with >95% purity and low endotoxin, it is suitable for biochemical assays, cell-based studies, and functional validation.
  • Lyophilized format for extended shelf life and easy transport.
  • N-terminal His tag enables affinity purification and detection.
  • High purity (>95%) supports reliable biochemical assays.
  • Low endotoxin (<1 EU/μg) minimizes confounding immune responses in cell assays.
  • Validated biological activity with a low ED50 in functional assays.
Catalog No. 50-001-85150