Learn More
ABclonal Technology S100A9 Rabbit PolymAb
SDP
Supplier:  ABclonal Technology A26782PM100UL
Encompass_Preferred
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs S100 proteins are localized in the cytoplasm and or nucleus of a wide range of cells and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation S100 genes include at least 13 members which are located as a cluster on chromosome 1q21 This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis This antimicrobial protein exhibits antifungal and antibacterial activity
Catalog No. 50-000-08563