Learn More
Cayman Chemical S100BCTerm RabbIt Monclo 100uL
SDP
Supplier:  Cayman Chemical 32249100 UL
Encompass_Preferred
An inhibitor of SIK2 (IC50 50s 21.63 and 6.63 nM respectively) inhibits TNF-a secretion in RAW 264.7 cells (IC50 ~2.5 UM) and reduces phosphorylation of CRTC3 and HDAC4 in macrophages at 3 UM inhibits growth of 10 ovarian cancer cell lines (IC50s 0.8-2.6 UM) and increases sensitivity of eight of them to paclitaxel halts the cell cycle at the G2/M phase and induces apoptosis and tetraploidy in SKOV3 cells at 1 UM has an additive effect on reducing tumor growth when used in combination with paclitaxel in an ovarian cancer mouse xenograft model at 60 mg/kg per day
Catalog No. 50-294-9305