Learn More
Genetex Inc S100P Monoclonal Antibody [MSVA-480R] HistoMAX™ (GTX04398) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04398
Encompass_Preferred
The S100P antibody [MSVA-480R] HistoMAX™ (GTX04398) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human muscle-invasive bladder cancer (MIBC) tissue using GTX04398 S100P antibody [MSVA-480R] HistoMAX. Muscle invasive urothelial carcinoma of the urinary bladder showing a strong S100P immunostaining of tumor cells.
Catalog No. NC3666763