Learn More
Medchemexpress LLC S100P Human N-His 5ug
SDP
Supplier:  Medchemexpress LLC HYP71138A5UG
Encompass_Preferred
Recombinant human S100P protein with an N-terminal His tag, supplied as a lyophilized powder for use in biochemical assays, protein-protein interaction studies, and antibody validation. Expressed in E. coli and characterized by SDS-PAGE for high purity; follow manufacturer recommendations for reconstitution and storage to preserve activity.
  • N-terminal His tag for affinity purification and detection.
  • High purity (>95%) for reliable assay performance.
  • Lyophilized format for extended shelf life and easy shipping.
  • Suitable for biochemical assays and antibody validation.
  • Expressed in E. coli for consistent recombinant yields.
  • Stable when aliquoted and stored at -20°C or -80°C.
Catalog No. 50-001-97243