Learn More
Medchemexpress LLC S100P-IN-1 | 690991-06-1 | 99.8% | 440.40 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY146695
Encompass_Preferred
S100P-IN-1 is an S100P inhibitor with an IC50 of 22.7 nM. It demonstrates anti-metastatic effects on pancreatic cancer cells by blocking S100P binding to RAGE in a dose-dependent manner.
  • Blocks S100P binding to RAGE in a dose-dependent manner.
  • Exhibits S100P-specific inhibitory effects on cell invasion.
  • Shows no significant cytotoxicity at 10 μM for 24 hours.
  • Significantly inhibits invasion in BxPC-3 cells.
Catalog No. 50-003-63980