Learn More
KAYAKU ADVANCED MATERIALS INC S1813 Positive Photoresist, 1 QT, Red Amber liquid
Supplier:  KAYAKU ADVANCED MATERIALS INC 11134041
Encompass
KAYAKU ADVANCED MATERIALS S1813 Positive Photoresist is a high-performance positive photoresist designed for microelectronic device fabrication. This material is essential for creating precise patterns on substrates such as in semiconductor manufacturing and integrated circuit prototyping. The formulation is engineered with a toxicologically safer solvent, providing a reliable solution for photolithographic processes.
  • Ensures high resolution and pattern definition for nanoscale patterning.
  • Offers excellent coating properties with striation-free, uniform coatings.
  • Optimized for g-line exposure and effective with broad-band light sources.
  • Compatible with both metal-ion-free and metal-ion-bearing developers.
  • Dyed versions minimize notching and maintain linewidth control, enhancing process yield.
Catalog No. NC1165634