Learn More
Medchemexpress LLC S186 | 97759-16-5 | 99.0% | 321.15 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13950100MG
Encompass_Preferred
S186 is a new type of strontium-specific chelator. It is intended for research use only and is not for sale to patients.
  • Strontium-specific chelator
Catalog No. 50-003-51086