Learn More
Medchemexpress LLC S186 | 97759-16-5 | 99.0% | 321.15 | 50 MG

Supplier: Medchemexpress LLC HY1395050MG
S186 is a novel strontium-specific chelator designed exclusively for research applications. It presents as a white to off-white solid.
- Strontium-specific chelator
- For research use only
- White to off-white solid appearance
- Purity of 99.01%
- Molecular weight of 321.15
- Soluble in water at 25 mg/mL
- Insoluble or slightly soluble in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.