Learn More
American Radiolabeled Chemicals Inc S1P 33P A 10UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARP 0144A10UC
Encompass
Sphingosine, D-erythro-33P 1-phosphate 10uCi in 85% Methanol:15% water with 0.1% tetrabutyl ammonium hydroxide, 1 mCi/ml, 2000-3000 Ci/mmol, M.W. 379.5, Shipped in Dry Ice
Catalog No. 50-352-895
Only null left
Add to Cart
Request a Quote