Learn More
Waters Corp Waters Spherisorb ODS2 Column, 80Å Pore, 3 μm Particle, 4.6 mm × 100 mm
Supplier:  Waters Corp PSS832112
Encompass_Preferred
These general-purpose, silica-based, reversed-phase C18 columns feature intermediate ligand density and a specified carbon load of 11.5%. They are long-lasting, and high-efficiency chromatographic columns, offering flexibility in optimizing procedures and lowering solvent use. Manufactured under cGMP, ISO 9002 conditions, these columns ensure critical performance features like column efficiency, peak symmetry, column lifetime, and reproducibility. Their high-quality bonded phase allows for a wide range of separation selectivities.
  • Silica-based, reversed-phase C18 columns
  • Intermediate ligand density with a specified carbon load of 11.5%
  • Manufactured under cGMP, ISO 9002 conditions
  • Ensures high column efficiency, peak symmetry, and column-to-column reproducibility
  • High-quality bonded phase for a wide range of separation selectivities
Catalog No. 50-818-768