Learn More
Selleck Chemical LLC S55746 S8759-5MG
SDP
Supplier:  Selleck Chemical LLC S87595MG
Encompass_Preferred
S55746 (S 055746 BCL201) is a novel orally active BCL-2 specific inhibitor (Ki 1 3 nM) with poor affinity for BCL-XL and no significant binding to MCL-1 BFL-1 (BCL2A1/A1) The selectivity of S55746 for BCL-2 versus BCL-XL ranges from 70 to 400 folds
Catalog No. 50-000-24298