Learn More
Apexbio Technology LLC S63845, 5mg. Cas: 1799633-27-4 MFCD: MFCD31382374. MCL1 inhibitor
SDP
Supplier:  Apexbio Technology LLC A87375
Encompass_Preferred
S63845 is a small molecule MCL1 inhibitor with Ki < 1.2 nM [1].Myeloid cell leukemia 1 (MCL1) is a pro-survival protein and belongs to BCL-2 family proteins. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5182