Learn More
Medchemexpress LLC S65487 sulfate | 2416937-01-2 | 99.35% | 815.33 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY138697A10MM/1ML
Encompass_Preferred
S65487 sulfate is a potent and selective Bcl-2 inhibitor, functioning as a proagent of S55746. It induces apoptosis and exhibits anticancer activities by binding to the BH3 hydrophobic groove of Bcl-2.
  • Active against Bcl-2 mutations like G101V and D103Y
  • Induces apoptosis and inhibits cell proliferation in hematological cancer cell lines
  • Shows poor affinity with Mcl-1, Bfl-1, and Bcl-XL
  • Induces complete regression in Bcl-2-dependent RS4;11 tumors in vivo after a single intravenous administration
  • Demonstrates strong and persistent tumor regression in xenograft models of lymphoid malignancies when combined with a Mcl-1-specific inhibitor
Catalog No. 50-003-51245