Learn More
Sino Biological Peptide Substrates: S6K Substrate
Supplier:  Sino Biological S0558
Encompass_Preferred
The S6K sub peptide sequence (KRRRLASLR) is based on human 40S ribosomal protein S6 (amino acid 230-238).
Catalog No. 50-256-7903