Learn More
Cayman Chemical S7 Nuclease asy Reagent 50uL
SDP
Supplier:  Cayman Chemical 60101150 UL
Encompass_Preferred
Source Recombinant SARS-CoV-2 N-Terminal His-tagged nsp1 expressed in E. coli Amino acids 13-128
Catalog No. 50-289-6597