Learn More
Electron Microscopy Sciences PS78 Micrometer Standard with NPL Cert
SDP
Supplier:  Electron Microscopy Sciences 6804778NP
Encompass_Preferred
These stage micrometers are available with a calibration certificate that provides traceability for the precise calibration and confirmation of accuracy of optical measuring instruments, necessary under ISO provisions. The glass discs are mounted in stainless steel slides with engraved serial numbers. Each slide is supplied in a polished wooden storage case. Calibrated by the National Physical Laboratory by taking measurements at selected points on the scale.
Catalog No. 50-290-05