Learn More
MOLTOX Molecular Toxicology Inc Sprague-Dawley Rat Liver S9, Male, Uninduced, 0.154M KCL, 2 mL/vial, Store in Ultra-low
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 11102.2
Encompass
Uninduced Sprague Dawley Rat Liver S9. For use in control experiments. This S9 is prepared with untreated tissue. It is imperative that frozen S9 preparations remain in storage conditions of around -80C, requiring expedited shipping on dry ice and careful temperature considerations during the thaw process. Once thawed your S9 is ready for use with cofactors. MOLTOX S9 preparations and cofactor reagents for use in metabolic activation studies are manufactured to rigorous standards of quality and performance and are used by leading industrial, government and academic laboratories worldwide. *Non Returnable*
Catalog No. 50-948-576