Learn More
Medchemexpress LLC S961 acetate | 99.83% | 4802.12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYP2093B50MG
Encompass_Preferred
S961 acetate is a high-affinity and selective antagonist of the insulin receptor (IR). It exhibits IC50 values of 0.048 nM for HIR-A, 0.027 nM for HIR-B, and 630 nM for human insulin-like growth factor I receptor (HIGF-IR) in SPA-assay. It also demonstrates high-affinity for Rat IR and Pig IR with IC50s of 0.056 nM and 0.084 nM, respectively, in PEG-assay.
  • High-affinity and selective antagonist of the insulin receptor.
  • Targets HIR-A, HIR-B, HIGF-IR, Rat IR, and Pig IR.
Catalog No. 50-004-24142