Learn More
Genscript Corporation SAA ML95 mAb Mouse
Supplier:  Genscript Corporation V0200710
Encompass
Serum Amyloid A (SAA) is an acute-phase protein The concentration of SAA in blood increases rapidly when tissue damage or inflammation occurs It is considered as an early biomarker for the diagnosis of many inflammatory diseases SAA can also be used as an important indicator of infection
Catalog No. 50-000-57359