Learn More
Medchemexpress LLC Sabcomeline hydrochloride | 159912-58-0 | 99.8% | 229.71 | C10H16ClN3O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY106432A25MG
Encompass_Preferred
Sabcomeline hydrochloride is the hydrochloride salt of sabcomeline, a functionally selective muscarinic M1 receptor partial agonist used in neuroscience research. It is commonly used to investigate M1-mediated signaling and cognitive function in preclinical Alzheimer's disease models. The compound is provided as a characterized solid suitable for in vitro and in vivo studies.
Catalog No. 50-002-42086