Learn More
Medchemexpress LLC Saccharic acid | 87-73-0 | MFCD09752094 | 98.0% | 210.14 | C6H10O8 | 25 MG

Supplier: Medchemexpress LLC HYW34588525MG
Saccharic acid is a small-molecule research reagent that acts as a competitive inhibitor of β-glucuronidase and is used to retard hydrolysis of glucuronide substrates in tissue and enzymatic assays. It is provided as a high-purity compound for biochemical and histochemical studies.
- Competitive inhibitor of β-glucuronidase.
- Delays hydrolysis of glucuronide substrates in tissue sections.
- Purity 98.0%.
- CAS number 87-73-0.
- Chemical formula C6H10O8 and molecular weight 210.14.
- Pack size 25 MG suitable for laboratory research.
- Intended for research use in biochemical assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.