Learn More
Medchemexpress LLC Saccharic acid | 87-73-0 | MFCD09752094 | 98.0% | 210.14 g/mol | C6H10O8 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW345885100MG
Encompass_Preferred
Saccharic acid is a research-grade small molecule used as a competitive inhibitor of β-glucuronidase in biochemical and pharmacological studies. It is supplied as a purified, water-soluble solid intended for enzyme inhibition assays and tissue-based experiments.
  • Competitive β-glucuronidase inhibitor useful in enzyme activity and metabolism studies.
  • High purity (98.0%) solid suitable for biochemical assays.
  • Water soluble for convenient preparation of aqueous solutions.
  • Well-characterized compound with CAS number 87-73-0 for unambiguous identification.
  • Molecular weight 210.14 g/mol for stoichiometric calculations.
Catalog No. 50-002-80922