Learn More
Volu Sol Saccomanno Solution, 32 oz / 1 L, Volu-Sol™
SDP
Supplier:  Volu Sol VMA032
Encompass_Preferred
Premixed fixative, used in collecting mucoid specimens for cytologic diagnosis
Catalog No. NC9183554