Learn More
Medchemexpress LLC Sacituzumab govitecan 5mg
SDP
Supplier:  Medchemexpress LLC HY1322545MG
Encompass_Preferred
Sacituzumab govitecan (IMMU-132) is an antibody-drug conjugate (ADC) targeting Trop-2 for delivery of SN-38. Sacituzumab govitecan shows anticancer activity, 1491917-83-9, 5mg
Catalog No. NC2187566