Learn More
Medchemexpress LLC Sacubitril (AHU-377) | 149709-62-6 | 99.9% | 411.49 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHY15407
Encompass_Preferred
Sacubitril (AHU-377) is a potent, orally active neprilysin (NEP) inhibitor with an IC50 of 5 nM. It is a component of the heart failure medicine LCZ696 and is used for the research of heart failure, hypertension, and COVID-19. It enhances the tone of the natriuretic peptide (NP) system and exerts significant antihypertensive effects.
  • Potent and orally active neprilysin (NEP) inhibitor
  • IC50 of 5 nM for NEP
  • Component of heart failure medicine LCZ696
  • Used for research on heart failure, hypertension, and COVID-19
  • Enhances natriuretic peptide (NP) system tone
  • Exerts significant antihypertensive effects
  • Converted to active metabolite LBQ657
  • Active metabolite inhibits cardiac myocyte hypertrophy
  • Potentiates ANF-induced urinary sodium excretion
  • Augments ANP-evoked plasma cGMP levels
Catalog No. 50-003-64865