Learn More
Selleck Chemical LLC Sacubitril/valsartan
SDP
Supplier:  Selleck Chemical LLC S767810MM/1ML
Encompass_Preferred
Sacubitril/valsartan (LCZ696 Sacubitril Valsartan) consisting of valsartan and sacubitril in 1 1 molar ratio is an orally bioavailable dual-acting angiotensin receptor-neprilysin inhibitor (ARNi) for hypertension and heart failure Phase 3
Catalog No. 50-000-00119