Learn More
Medchemexpress LLC Sacubitril/Valsartan | 936623-90-4 | 100.0% | 957.99 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY18204A500MG
Encompass_Preferred
Sacubitril/Valsartan (LCZ696) is comprised of Valsartan and Sacubitril (AHU377) in a 1:1 molar ratio. It is a first-in-class, orally bioavailable, and dual-acting angiotensin receptor-neprilysin (ARN) inhibitor. It is used for hypertension and heart failure and ameliorates diabetic cardiomyopathy by inhibiting inflammation, oxidative stress, and apoptosis.
  • First-in-class and orally bioavailable.
  • Dual-acting angiotensin receptor-neprilysin (ARN) inhibitor.
  • Ameliorates diabetic cardiomyopathy by inhibiting inflammation, oxidative stress, and apoptosis.
  • Exhibits small weights and reduces interstitial fibrosis in cardiac remodeling and dysfunction after myocardial infarction.
Catalog No. 50-003-87684