Learn More
Medchemexpress LLC Sacubitril/Valsartan | 936623-90-4 | 99.99% | 957.99 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY18204A10MM/1ML
Encompass_Preferred
Sacubitril/Valsartan, also known as LCZ696, is a first-in-class, orally bioavailable, dual-acting angiotensin receptor-neprilysin (ARN) inhibitor. Comprised of Valsartan and Sacubitril (AHU377) in a 1:1 molar ratio, it is used in research for hypertension and heart failure. This compound has been shown to improve diabetic cardiomyopathy by inhibiting inflammation, oxidative stress, and apoptosis.
  • First-in-class, orally bioavailable, dual-acting inhibitor
  • Ameliorates diabetic cardiomyopathy
  • Inhibits inflammation, oxidative stress, and apoptosis
Catalog No. 50-003-87668