Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sacubitrilat impurity 34 | 149709-58-0 | 99.68% | C20H23NO3 | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHY78834

Encompass_Preferred

Sacubitrilat impurity 34 is an impurity of Sacubitrilat. It is intended for research use only.

  • Purity: 99.68%
  • Molecular weight: 325.41
  • Formula: C20H23NO3
  • CAS No.: 149709-58-0
  • Appearance: Solid
  • Color: White to yellow

Catalog No. 50-003-58695


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.