Learn More
Chemglass Life Sciences Saddle O-ring ONLY (replacement part)
SDP
Supplier:  Chemglass Life Sciences CG207720
Encompass_Preferred
Saddle o-ring replacement for the CG2077 series of stirrer bearings. The saddle o-ring provides only glass and PTFE contact with the reaction, while allowing for the vacuum tight seal. Design of the bearing permits replacement of the saddle o-ring at a fraction of the cost of the entire bearing.
Catalog No. CG207720