Learn More
Lonza Walkersville SAEC - for S-ALI Medium
Supplier:  Lonza Walkersville CC2547S
Encompass
Lonza s Human Small Airway Epithelial Cells (SAEC) are isolated from normal human lung tissue the distal portion of the lung in the 1mm bronchiole area Contact our Scientific Support team for additional details Cryopreserved Clonetics SAECs are guaranteed through 15 population doublings and stain positive for cytokeratin 19 SAECs labeled CC-2547S are suitable for air-liquid interface studies and are guaranteed for differentiation marked by cilia formation mucin production andTEER levels All cells test negative for mycoplasma bacteria yeast and fungi HIV-1 hepatitis B andhepatitis C are not detected for all donors and/or cell lots A Certificate of Analysis is provided for each cell lot purchased
Catalog No. 50-004-96398