Learn More
Research Products International Corp Safety-Solve Complete Counting Cocktail, 4 Liters
SDP
Supplier:  Research Products International Corp 111177
Encompass_Preferred
Safety-Solve is a ready-to-use high flash point, cocktail for liquid scintillation counting. This low-odor, high flash point, low viscosity formula does not require dilution prior to use. Contains a blend of high quantum efficiency quench resistant fluors in a non-ionic emulsifier system which will accommodate a wide range of samples.
Catalog No. 50-212-986