Learn More
Medchemexpress LLC Safinamide mesylate | 202825-46-5 | 99.1% | C18H23FN2O5S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY70057A100MG
Encompass_Preferred
Safinamide mesylate is a potent, selective, and reversible monoamine oxidase B (MAO-B) inhibitor. It also blocks sodium channels and modulates glutamate release. It exhibits neuroprotective and neurorescuing effects and can be used for studies related to Parkinson's disease and ischemic stroke.
  • Potent, selective, and reversible monoamine oxidase B inhibitor
  • Blocks sodium channels
  • Modulates glutamate release
  • Exhibits neuroprotective and neurorescuing effects
  • Useful for studies related to Parkinson's disease
  • Useful for studies related to ischemic stroke
Catalog No. 50-004-34811