Learn More
TARGETMOL CHEMICALS INC SAFIT2 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T168365MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SAFit2 is a highly effective and selective fk506 binding protein 51(FKBP51) inhibitor (Ki 6 nM). SAFit2 can enhance AKT2-AS160 binding and participate in the downstream response of glucocorticoid release in vivo and is a candidate compound for the treatment of obesity chronic pain depression and anxiety. purity: 99%
Catalog No. 50-258-1147