Learn More
Volu Sol Safranin O Stain, 128 oz / 3.78 L, Volu-Sol™
SDP
Supplier:  Volu Sol VGC128
Encompass_Preferred
Safranin O, component of the Gram Stain Kit.  General biological stain used in histology and cytology.  Safranin colors cell nuclei red, making it a common counterstain in some staining protocols
Catalog No. 50-420-807