Learn More
Medchemexpress LLC SAG-524 | 2246696-89-7 | 98.5% | 594.12 g·mol⁻1 | C30H32ClN5O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15799310MG
Encompass_Preferred
SAG-524 is a novel, orally bioavailable small-molecule inhibitor of hepatitis B virus (HBV) replication that destabilizes HBV RNA. In cell-based assays it reduced extracellular HBV-DNA and HBsAg levels with IC50 values of 0.92 nM and 1.4 nM, respectively. It is supplied as a high-purity research solid for in vitro studies.
  • Potent HBV RNA destabilizer reducing HBV-DNA and HBsAg (IC50 0.92 nM and 1.4 nM).
  • Orally bioavailable small molecule suitable for preclinical research.
  • High purity (98.5%).
  • Provided as a 10 mg solid for research use.
  • Key identifiers: CAS 2246696-89-7; molecular formula C30H32ClN5O4S; molecular weight 594.12 g·mol⁻1.
  • Not intended for human or veterinary use; for laboratory research only.
Catalog No. 50-002-88424