Learn More
AdipoGen SAG
SDP
Supplier:  AdipoGen AGCR13506M025
Encompass_Preferred
Chemical. CAS 912545-86-9. Formula C28H28ClN3OS. MW 490.1. Cell permeable Sonic hedgehog Shh agonist. Cell permeable smoothened Smo agonist. Induces Sonic hedgehog Shh pathway activation and counteracts cyclopamine inhibition of Smo. Acts as an activator of Smo at low concentrations and as an inhibitor of Smo at very high concentrations. Potent activator of VEGF expression in vitro.
Catalog No. 50-256-2340